(R)-(+)-1,2-Epoxypropane 98+%

Danger

Synonyms: (R)-(+)-Propene oxide(R)-(+)-Propylene oxide(R)-(+)-1,2-Propylene oxide(R)-(+)-Methyloxirane

AC39957-0250 AC39957-0050
102777-026EA 284 USD
102777-026 102775-988
(R)-(+)-1,2-Epoxypropane 98+%
(R)-(+)-1,2-Epoxypropane
Formula: C₃H₆O
MW: 58.08 g/mol
Melting Pt: –112 °C
Density: 0.829 g/cm³
Flash Pt: –37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066211
CAS Number: 15448-47-2
UN: 1280
ADR: 3,I

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3680 (20 °C, 589 nm)
GC ≥98.0 %
Specific optical rotation +12° to +15° (20 °C, 589 nm) (Neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR