2-Chloroquinoline-3-carbaldehyde 98%

Warning

AC39955-0100 AC39955-0010
102774-074EA 30.8 USD
102774-074 102776-134
2-Chloroquinoline-3-carbaldehyde 98%
2-Chloroquinoline-3-carbaldehyde

Appearance: Yellow to yellow-brown Crystalline powder

Formula: C₁₀H₆ClNO
MW: 191.62 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00130079
CAS Number: 73568-25-9

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR