4-Thioanisolemagnesium bromide 0.5 M in THF, AcroSeal™

Danger

AC39907-1000
102777-076EA 273 USD
102777-076
4-Thioanisolemagnesium bromide 0.5 M in THF, AcroSeal™
4-Thioanisolemagnesium bromide
Formula: C₇H₇BrMgS
Flash Pt: <–17 °C
Storage Temperature: Ambient
MDL Number: MFCD00672010
CAS Number: 18620-04-7
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.4 to 0.6 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR