ω-Bromo-2-nitroacetophenone 98%

Danger

Synonyms: 2-Bromo-2'-nitroacetophenone 2-Nitrophenacyl bromide Bromomethyl o-nitrophenyl ketone α-Bromo-2-nitroacetophenone

AC39885-0050 AC39885-0010
102776-334EA 169 USD
102776-334 102774-610
ω-Bromo-2-nitroacetophenone 98%
ω-Bromo-2-nitroacetophenone
Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00010294
CAS Number: 6851-99-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR