2,5-Difluorobenzenesulfonyl chloride 97%

Danger

Synonyms: 2,5-Difluorobenzene-1-sulfonyl chloride2,5-Difluorobenzene-1-sulphonyl chloride

AC39879-0050 AC39879-0010
102775-090EA 80.2 USD
102775-090 102776-686
2,5-Difluorobenzenesulfonyl chloride 97%
2,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060682
CAS Number: 26120-86-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR