2-Benzyloxyaniline 97%

Warning

Synonyms: 2-(Benzyloxy)aniline

AC39830-0010
102773-984EA 27.9 USD
102773-984
2-Benzyloxyaniline 97%
2-Benzyloxyaniline
Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD03093873
CAS Number: 20012-63-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to pink to brown Low melting solid
Infrared spectrum Conforms
Melting point 35 to 39 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR