(R)-(+)-1-Boc-3-aminopyrrolidine 97%

Danger

Synonyms: (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate(3R)-(+)-1-Boc-3-aminopyrrolidine(3R)-(+)-3-Amino-1-Boc-pyrrolidine(3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine

AC39827-0050 AC39827-0010
102777-486EA 411 USD
102777-486 102775-964
(R)-(+)-1-Boc-3-aminopyrrolidine 97%
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419272
CAS Number: 147081-49-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +2° to +4° (20 °C, 589 nm) (c=1 in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR