2-Chloro-5-formylpyridine 97%

Warning

Synonyms: 6-Chloronicotinaldehyde6-Chloropyridine-3-carboxaldehyde2-Chloropyridine-5-carboxaldehyde

AC39810-0050 AC39810-0010
102775-618EA 100 USD
102775-618 102777-250
2-Chloro-5-formylpyridine 97%
2-Chloro-5-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03095223
CAS Number: 23100-12-1

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 82 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR