2-Fluoro-5-(trifluoromethyl)phenol 98%

Warning

Synonyms: 1-Fluor-2-hydroxy-4-(trifluormethyl)benzol2-Fluoro-5-trifluoromethylphenol

AC39784-0010
102774-096EA 27.5 USD
102774-096
2-Fluoro-5-(trifluoromethyl)phenol 98%
2-Fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 146 °C (1013 hPa)
Density: 1.436 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR