2-Bromo-5-fluoropyridine 98%

Warning

AC39797-0010 AC39797-0050
102775-534EA 114 USD
102775-534 102774-028
2-Bromo-5-fluoropyridine 98%
2-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 83 °C (44 mmHg)
Melting Pt: 30…31 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless white to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR