Sarcosine methyl ester hydrochloride 97%

Warning

Synonyms: H-Sar-OMe·HClMethyl N-methylglycinate hydrochloride

AC39750-2500 AC39750-0500
102777-444EA 319 USD
102777-444 102775-828
Sarcosine methyl ester hydrochloride 97%
Sarcosine methyl ester hydrochloride
Formula: C₄H₉NO₂·HCl
MW: 139.58 g/mol
Melting Pt: 110…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00038876
CAS Number: 13515-93-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1.0 % (1 g, 70 °C) (to constant weight)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR