3-Bromobenzoyl chloride 97%

Danger

Synonyms: m-Bromobenzoyl chloride3-Bromobenzoylchloride

AC39745-1000 AC39745-0250 AC39745-0050
102777-202EA 340 USD
102777-202 102775-580 102774-060
3-Bromobenzoyl chloride 97%
3-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 74.5…75 °C (0.5 mmHg)
Density: 1.666 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5940 to 1.5960 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR