2-Amino-5-fluoropyridine 95%

Warning

AC39729-0050 AC39729-0010
102775-456EA 72.7 USD
102775-456 102777-088
2-Amino-5-fluoropyridine 95%
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
Melting Pt: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD01861120
CAS Number: 21717-96-4
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 93 to 99 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR