Cyclopentyl methyl ether (CPME), extra dry 99.5% stabilized, AcroSeal™
|
Danger
|
AC39725-1000
102774-530EA
36
USD
102774-530
Cyclopentyl methyl ether (CPME), extra dry 99.5% stabilized, AcroSeal™
Cyclopentyl methyl ether (CPME)
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 106 °C (760 mmHg) Melting Pt: –140 °C Density: 0.86 g/cm³ Flash Pt: –1 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276401 CAS Number: 5614-37-9 UN: 3271 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.4 % |
Stabilizer | Approx. 50 ppm BHT |
Peroxide value | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...