Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal™

Danger

Synonyms: Ethylene glycol monomethyl ether

AC39689-0010 AC39689-1000
102774-706EA 45.12 USD
102774-706 103528-472
Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal™
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124 °C (760 mmHg)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR