1H-Pyrazole-4-carboxylic acid 97%

Warning

Synonyms: 4-Pyrazolecarboxylic acid

AC39685-5000 AC39685-1000
102776-174EA 144 USD
102776-174 102774-836
1H-Pyrazole-4-carboxylic acid 97%
1H-Pyrazole-4-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 282 °C
Storage Temperature: Ambient
MDL Number: MFCD00011558
CAS Number: 37718-11-9

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR