(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilized, AcroSeal™

Danger

AC39663-0010 AC39663-1000
102775-992EA 167 USD
102775-992 102775-086
(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilized, AcroSeal™
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: –136 °C
Density: 0.86 g/cm³
Flash Pt: –11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005367
CAS Number: 96-47-9
UN: 2536
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.002 %
GC ≥99.0 %
Stabilizer 150 to 400 ppm BHT
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR