Allylboronic acid pinacol ester 98%

Warning

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

AC39513-0050 AC39513-0010
102774-124EA 40.1 USD
102774-124 102775-860
Allylboronic acid pinacol ester 98%
Allylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
MW: 168.05 g/mol
Boiling Pt: 50…53 °C (5 mmHg)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR