2-Bromo-3-nitrotoluene 98%

Warning

AC39410-0050 AC39410-0010
102774-660EA 46 USD
102774-660 102775-968
2-Bromo-3-nitrotoluene 98%
2-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 135…136 °C (8 mmHg)
Melting Pt: 38…40 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134555
CAS Number: 41085-43-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals or granules and/or chunks
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR