4-Methoxypiperidine 98+%

Danger

AC39339-0010 AC39339-0050
102776-166EA 136 USD
102776-166 102777-460
4-Methoxypiperidine 98+%
4-Methoxypiperidine
Formula: C₆H₁₄ClNO
MW: 115.1700 g/mol
Boiling Pt: 44 °C (9mbar)
Storage Temperature: Ambient
MDL Number: MFCD04115010
CAS Number: 4045-24-3
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR