4-(Chloromethyl)benzoic acid 98%

Danger

AC39289-0500
102777-198EA 278 USD
102777-198
4-(Chloromethyl)benzoic acid 98%
4-(Chloromethyl)benzoic acid
Formula: C₈H₇ClO₂
MW: 170.60 g/mol
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00002568
CAS Number: 1642-81-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR