1-Bromoisoquinoline 95+%

Danger

AC39259-0010 AC39259-0050
102777-094EA 345 USD
102777-094 102775-590
1-Bromoisoquinoline 95+%
1-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.0546 g/mol
Melting Pt: 43…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00234478
CAS Number: 1532-71-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 42 to 48 °C
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR