(3-Chloropropyl)triethoxysilane 97+%

Synonyms: 3-(Triethoxysilyl)propylchloride3-ChloropropyltriethoxysilaneTriethoxy(3-chloropropyl)silane

AC39189-0010
103514-144EA 150 USD
103514-144
(3-Chloropropyl)triethoxysilane 97+%
(3-Chloropropyl)triethoxysilane
Formula: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
MW: 240.8 g/mol
Boiling Pt: 230 °C (760 mmHg)
Density: 1.004 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00018985
CAS Number: 5089-70-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20 °C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR