2-Iodobenzonitrile 99%

Warning

Synonyms: o-Iodobenzonitrile

AC39180-0050 AC39180-0250 AC39180-0010
102775-192EA 81.4 USD
102775-192 102773-994 102777-082
2-Iodobenzonitrile 99%
2-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Melting Pt: 54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00079761
CAS Number: 4387-36-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Solid
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR