α-Bromo-4-chlorotoulene 98%

Danger

Synonyms: alpha-Bromo-4-chlorotoluene4-Chlorobenzyl bromide1-(Bromomethyl)-4-chlorobenzene

AC39171-0050 AC39171-0250 AC39171-1000
102775-976EA 165 USD
102775-976 102777-502 102774-666
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 118…123 °C (19 mmHg)
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00040714
CAS Number: 622-95-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to almost white Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR