1,4-Diethynylbenzene 95%

Warning

Synonyms: p-Diethynylbenzene1,4-Diethylnylbenzene

AC39155-0050 AC39155-0010
102775-978EA 140 USD
102775-978 102774-668
1,4-Diethynylbenzene 95%
1,4-Diethynylbenzene
Formula: C₁₀H₆
MW: 126.16 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078375
CAS Number: 935-14-8

Order Now

Specification Test Results

Appearance White to orange to brown Crystalline powder, crystals and/or chunks
Infrared spectrum Conforms
Melting point 90 to 96 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR