3-Aminobenzylamine 97%

Danger

Synonyms: 3-(Aminomethyl)anilinealpha-Amino-m-toluidine

AC39144-0050 AC39144-0250
102775-954EA 203 USD
102775-954 102774-416
3-Aminobenzylamine 97%
3-Aminobenzylamine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Melting Pt: 36…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00078355
CAS Number: 4403-70-7
EINECS: 412-230-2
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light brown Low melting solid or liquid
Infrared spectrum Conforms
Melting point 38 to 44 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR