1-Chloroisoquinoline 97+%

Warning

AC39138-0050 AC39138-0010
102775-642EA 117 USD
102775-642 102774-092
1-Chloroisoquinoline 97+%
1-Chloroisoquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Melting Pt: 31…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00024134
CAS Number: 19493-44-8
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystals and/or chunks or low melting solid
Infrared spectrum Conforms
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR