Methyl-4-(chloromethyl)benzoate 97%

Danger

Synonyms: Methyl 4-(chloromethyl)benzoate

AC39131-0010
102773-958EA 29.3 USD
102773-958
Methyl-4-(chloromethyl)benzoate 97%
Methyl 4-(chloromethyl)benzoate
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 160…165 °C (30 mmHg)
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00511224
CAS Number: 34040-64-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Low melting solid
Infrared spectrum Conforms
Melting point 37 to 40 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR