4-(Chloromethyl)benzonitrile 98%

Danger

AC39115-0500 AC39115-0100
102776-786EA 219 USD
102776-786 102775-272
4-(Chloromethyl)benzonitrile 98%
4-(Chloromethyl)benzonitrile
Formula: C₈H₆ClN
MW: 151.60 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00019761
CAS Number: 874-86-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 80 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR