4-tert-Butylbenzylamine 97%

Danger

Synonyms: p-tert-Butylbenzylamine

AC39101-0010 AC39101-0050
102774-808EA 131 USD
102774-808 102773-778
4-tert-Butylbenzylamine 97%
4-tert-Butylbenzylamine
Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00040754
CAS Number: 39895-55-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR