(Dimethylamino)acetaldehyde dimethylacetal 97%

Danger

Synonyms: N-(2,2-Dimethoxyethyl)dimethylamine2,2-Dimethoxy-N,N-dimethylethanamine

AC39091-0050 AC39091-0010
102775-344EA 80.7 USD
102775-344 102773-920
(Dimethylamino)acetaldehyde dimethylacetal 97%
(Dimethylamino)acetaldehyde dimethylacetal
Formula: C₆H₁₅NO₂
MW: 133.1888 g/mol
Boiling Pt: 64…66 °C (12 mmHg)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00010205
CAS Number: 38711-20-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4120 (20 °C, 589 nm)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR