Sodium borohydride 12% in sodium hydroxide solution 40%

Danger

Synonyms: Sodium tetrahydridoborateSodium tetrahydroborate

AC38993-0010 AC38993-0025 AC38993-2500 AC38993-1000
102775-048EA 103.15 USD
102775-048 102774-376 102776-590 102773-878
Sodium borohydride 12% in sodium hydroxide solution 40%
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 3320
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless Liquid
Titration Iodometric 11 to 13 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR