Potassium sulfide (K2Sx), pure

Danger

Synonyms: Liver of sulfurSulfurated potashLiver of sulphurSulphurated potashPotassium polysulfidePotassium polysulphide

AC38959-0025 AC38959-2500
103528-370EA 229 USD
103528-370 102774-704
Potassium sulfide (K2Sx), pure
Potassium sulphide (K2Sx)
Formula: K₂(S)n
MW: 110.26 g/mol
Melting Pt: 200…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00148074
CAS Number: 37199-66-9
UN: 1382
ADR: 4.2,II

Order Now

Specification Test Results

Appearance Ochre to brown Lumps
Titration Iodometric 42.0 to 44.0 % (K2S)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR