Benzylmagnesium chloride 1.4 M in THF, AcroSeal™

Danger

AC38955-8000
102777-106EA 330.8 USD
102777-106
Benzylmagnesium chloride 1.4 M in THF, AcroSeal™
Benzylmagnesium chloride
Formula: C₇H₇ClMg
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000469
EINECS: 230-039-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear green-brown to brown-purple Solution (May develop some turbidity or precipitate)
Titration Grignard 19 to 21 % (As BMC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR