N-Boc-ethylenediamine 98%

Danger

Synonyms: t-boc-N-EDAtert-Butyl N-(2-Aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-ethylenediaminetert-Butyl (2-aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-diaminoethaneN-BOC-1,2-diaminoethane

AC38990-0050 AC38990-0250 AC38990-0010
102775-356EA 76 USD
102775-356 102776-978 102777-890
N-Boc-ethylenediamine 98%
N-Boc-ethylenediamine
Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 80…84 °C (.3 mmHg)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR