Benzyl-2-bromoethyl ether 97%

Warning

Synonyms: (2-Bromoethoxymethyl)benzene((2-Bromoethoxy)methyl)benzeneBenzyloxyethyl Bromide

AC38887-0010 AC38887-1000 AC38887-0250 AC38887-0050
102777-398EA 386 USD
102777-398 103528-334 102775-892 102774-106
Benzyl-2-bromoethyl ether 97%
Benzyl-2-bromoethyl ether

Appearance: Clear colorless to light yellow Liquid

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 254 °C (760 mmHg)
Density: 1.36 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD01321307
CAS Number: 1462-37-9
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR