Isovaline 97%

Synonyms: D-IsovalineD(-)-Isovaline

AC38803-2500
200055-404EA 272 USD
200055-404
Isovaline 97%
Isovaline

Appearance: White Crystalline powder or needles

Formula: C₅H₁₁NO₂
MW: 117.148 g/mol
Melting Pt: 276…278 °C
Storage Temperature: Ambient
MDL Number: MFCD00800497
CAS Number: 3059-97-0

Order Now

Specification Test Results

Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Specific optical rotation -9.0° to -12.0° (20 °C, 589 nm) (C=1, water)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR