Acetyl chloride 1 M in dichloromethane, AcroSeal™

Danger

AC38774-8000 AC38774-1000
200063-884EA 386.54 USD
200063-884 200063-882
Acetyl chloride 1 M in dichloromethane, AcroSeal™
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Argentometric 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR