2-Bromo-6-formylpyridine 97%

Warning

Synonyms: 6-Bromopyridine-2-carboxaldehyde6-Bromopyridine-2-carbaldehyde6-Bromo-2-pyridinecarboxaldehyde

AC38742-0050 AC38742-0010
102775-942EA 112 USD
102775-942 102774-378
2-Bromo-6-formylpyridine 97%
2-Bromo-6-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683546
CAS Number: 34160-40-2

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR