Benzyl dimethylphosphonoacetate 98%

Warning

Synonyms: Benzyl 2-(dimethoxyphosphoryl)acetateDimethyl (Benzyloxycarbonylmethyl)phosphonateDimethylphosphonoacetic Acid Benzyl EsterDimethyl (Cbz-methyl)phosphonate

AC38737-0250 AC38737-0050
200055-156EA 487 USD
200055-156 200055-158
Benzyl dimethylphosphonoacetate 98%
Benzyl dimethylphosphonoacetate

Appearance: Clear colorless to light brown Liquid

Formula: C₁₁H₁₅O₅P
MW: 258.21 g/mol
Boiling Pt: 148…150 °C (.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01872507
CAS Number: 57443-18-2

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20 °C, 589 nm)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR