2-Octyn-1-ol 98%

Warning

AC38703-0050 AC38703-0250
102775-758EA 137 USD
102775-758 102774-036
2-Octyn-1-ol 98%
2-Octyn-1-ol
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 76…78 °C (2 mmHg)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00039542
CAS Number: 20739-58-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR