Choline hydroxide 45% (w/w) in aqueous solution stabilized, pure

Danger

Synonyms: Choline2-Hydroxyethyltrimethylammonium Hydroxide

AC38672-0025 AC38672-1000 AC38672-0100 AC38672-5000
200055-340EA 72.2 USD
200055-340 200055-342 103528-258 103528-256
Choline hydroxide 45% (w/w) in aqueous solution stabilized, pure
Choline hydroxide
Formula: C₅H₁₅NO₂
MW: 121.18
Density: 1.07 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00002831
CAS Number: 123-41-1
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to tan Viscous solution
Stabilizer about 0.2 % ethylenediamine
Titration with HClO4 44.0 to 48.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR