2-Nitro-1-phenylpropene 99%

Warning

Synonyms: trans-β-Methyl-β-nitrostyrene

AC38651-0250 AC38651-0010 AC38651-0050
102773-720EA 20.6 USD
102773-720 102774-504 102776-370
2-Nitro-1-phenylpropene 99%
2-Nitro-1-phenylpropene
Formula: C₉H₉NO₂
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00014720
CAS Number: 705-60-2

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR