2-Amino-2-cyanoacetamide 97+%

Danger

AC38276-0050
200054-018EA 102 USD
200054-018
2-Amino-2-cyanoacetamide 97+%
2-Amino-2-cyanoacetamide
Formula: C₃H₅N₃O
MW: 99.0913 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00040532
EINECS: 229-771-3
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Beige to tan Powder
Infrared spectrum Conforms
Melting point 120 to 124 °C
Titration with HClO4 ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR