Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%

Warning

Synonyms: Bis(norbornadiene)rhodium(I) tetrafluoroborateBis(norbornadiene)rhodium (I) tetrafluoroborate

AC38255-5000
103532-802EA 239 USD
103532-802
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate 94%
Bis[eta-(2,5-norbornadiene)]rhodium(I) tetrafluoroborate

Appearance: Red Powder

Formula: C₁₄H₈BF₄Rh
MW: 365,92 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671775
CAS Number: 36620-11-8
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Red Powder
Infrared spectrum Conforms
Elemental Analysis ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR