Diethylaluminium iodide 0.9 M in toluene, AcroSeal™

Danger

AC38116-8000
103529-650EA 505 USD
103529-650
Diethylaluminium iodide 0.9 M in toluene, AcroSeal™
Diethylaluminium iodide
Formula: C₄H₁₀AlI
Storage Temperature: Ambient
MDL Number: MFCD00001025
CAS Number: 2040-00-8
EINECS: 218-032-0
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 19.5 to 20.5 wt % (on Al basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR