4-Phenyl-1-buten-4-ol 97%

Warning

Synonyms: 1-Phenyl-3-buten-1-ol

AC38086-0010 AC38086-0050
200054-754EA 27.4 USD
200054-754 200054-756
4-Phenyl-1-buten-4-ol 97%
4-Phenyl-1-buten-4-ol
Formula: C₁₀H₁₂O
MW: 148.20 g/mol
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00039617
CAS Number: 936-58-3

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5320 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR