Butyl-p-toluenesulfonate 99%

Warning

Synonyms: p-Toluenesulphonic acid Butyl Esterp-Toluenesulfonic acid Butyl EsterButyl 4-methylbenzenesulfonateButyl 4-methylbenzenesulphonate

AC38059-0250 AC38059-5000 AC38059-1000
200055-286EA 131 USD
200055-286 200055-288 103513-120
Butyl-p-toluenesulfonate 99%
Butyl-p-toluenesulfonate
Formula: C₁₁H₁₆O₃S
MW: 228.31 g/mol
Boiling Pt: 170…172 °C (10 mmHg)
Density: 1.12 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00027203
CAS Number: 778-28-9

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5050 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR