Methyl-5-iodsalicylate ≥99%

Warning

Synonyms: Methyl 2-hydroxy-5-iodobenzoate

AC38177-0100
200055-874EA 47.5 USD
200055-874
Methyl-5-iodsalicylate ≥99%
Methyl 5-iodsalicylate
Formula: C₈H₇IO₃
MW: 278.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016462
CAS Number: 4068-75-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 78 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR